Learn More
ATCC Proteus vulgaris
Supplier:  ATCC 8427
Encompass
Proteus vulgaris was isolated from an inner ear infection. This whole-genome sequenced bacterial strain can be used in media testing or as a quality control strain for BBL products.
Catalog No. 50-238-4268