Learn More
Hampton Research HR2-432/Proti-Ace 2 - 3 aliquots of 6 unique protease Pepsin, Thermolysin, Bromelain and Actinase E) and 6 aliquots of Proti-Ace Dilution Buffer
SDP
Catalog No. 502735
Encompass
HR2-432/Proti-Ace 2 - 3 aliquots of 6 unique proteases (Proteinase K, Clostripain (Endoproteinase-Arg-C), Pepsin, Thermolysin, Bromelain and Actinase E) and 6 aliquots of Proti-Ace Dilution Buffer. Please note items from this supplier are non-returnable.
Catalog No. 50-273-5 Supplier Hampton Research Supplier No. HR2432
HR2-432/Proti-Ace 2 - 3 aliquots of 6 unique proteases (Proteinase K, Clostripain (Endoproteinase-Arg-C), Pepsin, Thermolysin, Bromelain and Actinase E) and 6 aliquots of Proti-Ace Dilution Buffer. Please note items from this supplier are non-returnable.