Learn More
Cayman Chemical protin Carbonyl TCA SOLUTn 3mL
SDP
Catalog No. 502894321
Encompass_Preferred
A translocator protein ligand binds to translocator protein in a cell-free assay (Ki 3.6 nM) enhances pregnenolone synthesis in C6 rat glioma cells (EC50 10 nM)
Catalog No. 50-289-4321 Supplier Cayman Chemical Supplier No. 7015393 ML
A translocator protein ligand binds to translocator protein in a cell-free assay (Ki 3.6 nM) enhances pregnenolone synthesis in C6 rat glioma cells (EC50 10 nM)