Learn More
Cayman Chemical protin Carbonyl Wsh SOLUT 30mL
SDP
Catalog No. 502892322
Encompass_Preferred
Protein Carbonyl Wash Solution has been tested and formulated to work exclusively with Cayman’s Protein Carbonyl Fluorometric Assay Kit
Catalog No. 50-289-2322 Supplier Cayman Chemical Supplier No. 70153530 ML
Protein Carbonyl Wash Solution has been tested and formulated to work exclusively with Cayman’s Protein Carbonyl Fluorometric Assay Kit