Learn More
Apexbio Technology LLC Protease Inhibitor Cocktail(MS-SAFE, 50X in DMSO) 5x1ml
SDP
Supplier:  Apexbio Technology LLC K40015
Encompass_Preferred
Protease inhibitor cocktails prevent protein degradation during cell or tissue extraction These cocktails protect proteins from various proteolytic enzymes including serine cysteine acidic proteases and aminopeptidases that are present in cellular lysates This specific cocktail includes Aprotinin Bestatin E-64 and Leupeptin offering broad-spectrum inhibition against different protease types It is provided in a concentrated ready-to-use form in DMSO and is compatible with mass spectrometry (MS) analysis This cocktail does not contain AEBSF which can interfere with MS and EDTA is supplied separately for additional metalloprotease inhibition This product is ideal for biological and biomedical research especially when preserving protein samples for proteomics and functional protein studies
Catalog No. 50-000-63665