Learn More
Macherey-Nagel Protino Glutathione Agarose 4B, 10 mL
Catalog No. NC0389540
Encompass_Preferred
Protino Glutathione Agarose 4B (10 mL) Protino Glutathione Agarose 4B suspension for the purification of GST-tag proteins pack of 10 mL settled beaded agarose
Catalog No. NC0389540 Supplier Macherey-Nagel Supplier No. 745500.10
Protino Glutathione Agarose 4B (10 mL) Protino Glutathione Agarose 4B suspension for the purification of GST-tag proteins pack of 10 mL settled beaded agarose