Learn More
Macherey-Nagel Protino Ni-IDA Resin, 5 g
Catalog No. NC9597546
Encompass_Preferred
Protino Ni-IDA Resin (5 g) Protino Ni-IDA Resin for the purification of His-tag proteins pack of 5 g
Catalog No. NC9597546 Supplier Macherey-Nagel Supplier No. 745210.5
Protino Ni-IDA Resin (5 g) Protino Ni-IDA Resin for the purification of His-tag proteins pack of 5 g