Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Protionamide-d7 | 2468639-68-9 | 97.2% | 187.31 g/mol | C9H5D7N2S | 1 MG
SDP

Catalog No. 5000284032
Encompass_Preferred

Protionamide-d7 is a deuterium-labeled analogue of prothionamide provided as a small research sample for use in analytical and biological studies. It is intended for use as an isotopically labeled standard or tracer in mass spectrometry and metabolic investigations into mycobacterial activity.

  • Deuterium-labeled prothionamide for research applications.
  • Suitable as an internal standard or tracer in mass spectrometry.
  • CAS number 2468639-68-9 and formula C9H5D7N2S.
  • Molecular weight 187.31 g/mol.
  • Pack size 1 mg with reported purity approximately 97.2%.

Catalog No. 50-002-84032 Supplier Medchemexpress LLC Supplier No. HYW6543291MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Protionamide-d7 is a deuterium-labeled analogue of prothionamide provided as a small research sample for use in analytical and biological studies. It is intended for use as an isotopically labeled standard or tracer in mass spectrometry and metabolic investigations into mycobacterial activity.

  • Deuterium-labeled prothionamide for research applications.
  • Suitable as an internal standard or tracer in mass spectrometry.
  • CAS number 2468639-68-9 and formula C9H5D7N2S.
  • Molecular weight 187.31 g/mol.
  • Pack size 1 mg with reported purity approximately 97.2%.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.