Learn More
TARGETMOL CHEMICALS INC Protirelin Acetate 100MG
SDP
Catalog No. 502879354
Encompass_Preferred
Also available in 5 mg, 10 mg, 25 mg, 50 mg, 500 mg and bulk. Please contact Fisher for quotes. Protirelin Acetate (TRH Acetate) is a thyrotropin-releasing hormone, stimulating the release of thyrotropin and prolactin from the anterior pituitary. Purity 96.17%
Catalog No. 50-287-9354 Supplier TARGETMOL CHEMICALS INC Supplier No. TP1022100MG
Also available in 5 mg, 10 mg, 25 mg, 50 mg, 500 mg and bulk. Please contact Fisher for quotes. Protirelin Acetate (TRH Acetate) is a thyrotropin-releasing hormone, stimulating the release of thyrotropin and prolactin from the anterior pituitary. Purity 96.17%