Learn More
Medchemexpress LLC Protirelin acetate | 120876-23-5 | 99.9% | 452.46 | 1 ML
SDP
Catalog No. 5000425302
Encompass_Preferred
Protirelin acetate is a highly conserved neuropeptide that plays a crucial role in the hormonal regulation of thyroid-stimulating hormone (TSH) levels, in addition to exhibiting neuromodulatory functions. It is an evolutionarily ancient neuropeptide, with origins predating the divergence of protostomes and deuterostomes, and may have historically been involved in controlling postembryonic growth and reproduction.
  • Investigated for its effect on body temperature in rats.
  • Causes a temporary, dose-dependent rise in body temperature in male Wistar rats.
  • The hyperthermic action may involve the release of thyroid hormone.
  • Thyroidectomized rats do not show a rise in body temperature when administered Protirelin.
Catalog No. 50-004-25302 Supplier Medchemexpress LLC Supplier No. HYP0002A10MM/1ML
Protirelin acetate is a highly conserved neuropeptide that plays a crucial role in the hormonal regulation of thyroid-stimulating hormone (TSH) levels, in addition to exhibiting neuromodulatory functions. It is an evolutionarily ancient neuropeptide, with origins predating the divergence of protostomes and deuterostomes, and may have historically been involved in controlling postembryonic growth and reproduction.
  • Investigated for its effect on body temperature in rats.
  • Causes a temporary, dose-dependent rise in body temperature in male Wistar rats.
  • The hyperthermic action may involve the release of thyroid hormone.
  • Thyroidectomized rats do not show a rise in body temperature when administered Protirelin.