Learn More
Medchemexpress LLC Protirelin acetate | 120876-23-5 | CB8947811 | 99.9% | 452.46 | 50 MG
SDP
Catalog No. 5000426460
Encompass_Preferred
Protirelin acetate is a highly conserved neuropeptide that exerts hormonal control of thyroid-stimulating hormone (TSH) levels and also has neuromodulatory functions.
  • Exerts hormonal control of thyroid-stimulating hormone (TSH) levels
  • Functions as a neuromodulator
  • Targets the thyroid hormone receptor
  • Stable as powder for 2 years at -80°C or 1 year at -20°C
  • Stable in solvent for 6 months at -80°C or 1 month at -20°C
Catalog No. 50-004-26460 Supplier Medchemexpress LLC Supplier No. HYP0002A50MG
Protirelin acetate is a highly conserved neuropeptide that exerts hormonal control of thyroid-stimulating hormone (TSH) levels and also has neuromodulatory functions.
  • Exerts hormonal control of thyroid-stimulating hormone (TSH) levels
  • Functions as a neuromodulator
  • Targets the thyroid hormone receptor
  • Stable as powder for 2 years at -80°C or 1 year at -20°C
  • Stable in solvent for 6 months at -80°C or 1 month at -20°C