Learn More
Quality Biological Inc Protein Running Buffer 10X, 500 mL
SDP
Catalog No. 507517635
Encompass_Preferred
Protein Running Buffer, 10X is used as the electrophoresis buffer during the stacking and resolve process of sodium dodecyl sulfate ¿ polyacrylamide gel electrophoresis (SDS-PAGE), and is the running buffer for TRIS-glycine gel electrophoresis. ¿ Contains 1% SDS. ¿ Applications: This product is ideally suited for protein gel analysis in biochemical, molecular biological, cell biological, and immunological research studies. ¿ Storage Conditions: 15¿C to 30¿C ¿ Shipping Condition: ambient temperature ¿ Sterile Filtered ¿ DNase, RNase and Protease Tested ¿ Product and/or packaging contains recycled/renewable content ¿ Product and/or packaging is recyclable or reduces waste ¿ Product and/or packaging is safer to human and environmental health ¿ ¿ Max Shelf Life: 36 months
Catalog No. 50-751-7635 Supplier Quality Biological Inc Supplier No. 351132101
Add to Cart
Add to Cart
Protein Running Buffer, 10X is used as the electrophoresis buffer during the stacking and resolve process of sodium dodecyl sulfate ¿ polyacrylamide gel electrophoresis (SDS-PAGE), and is the running buffer for TRIS-glycine gel electrophoresis. ¿ Contains 1% SDS. ¿ Applications: This product is ideally suited for protein gel analysis in biochemical, molecular biological, cell biological, and immunological research studies. ¿ Storage Conditions: 15¿C to 30¿C ¿ Shipping Condition: ambient temperature ¿ Sterile Filtered ¿ DNase, RNase and Protease Tested ¿ Product and/or packaging contains recycled/renewable content ¿ Product and/or packaging is recyclable or reduces waste ¿ Product and/or packaging is safer to human and environmental health ¿ ¿ Max Shelf Life: 36 months