Learn More
MSC Proto Hex Bit Socket, 1/4 in, 1/16 in, 1 ea
Catalog No. 501920256
Encompass
This hand hex bit socket is designed for use with a 1/4 in drive, featuring a 1/16 in hex bit. Constructed from durable chrome alloy steel, it is suitable for various fastening applications.
  • Hex bit socket type
  • Drive size: 1/4 in
  • Hex size: 1/16 in
  • Bit length: 1 in
  • Chrome alloy steel material
  • Black oxide (stem) and polished chrome finish
  • Not tether capable
  • Non-sparking
  • Not insulated
Catalog No. 50-192-0256 Supplier MSC Supplier No. 39937578
This hand hex bit socket is designed for use with a 1/4 in drive, featuring a 1/16 in hex bit. Constructed from durable chrome alloy steel, it is suitable for various fastening applications.
  • Hex bit socket type
  • Drive size: 1/4 in
  • Hex size: 1/16 in
  • Bit length: 1 in
  • Chrome alloy steel material
  • Black oxide (stem) and polished chrome finish
  • Not tether capable
  • Non-sparking
  • Not insulated