Learn More
MSC Proto Hex Bit Socket, 1/4 in, 3 mm, 1 ea
Catalog No. 501920269
Encompass
The hex bit socket is designed for secure fastening and loosening tasks, featuring a 1/4 in drive for compatibility with various ratchets and drivers. Its 3 mm hex bit ensures precise fit for corresponding fasteners, making it suitable for a range of professional applications.
  • Hex opening securely holds the bit for optimal torque transfer
  • Constructed from heat-treated alloy steel for durability
  • Features a polished chrome finish for corrosion resistance
  • Bit length of 1 in for versatile use
  • Overall length of 1-7/8 in
  • Not insulated
  • Non-sparking
Catalog No. 50-192-0269 Supplier MSC Supplier No. 39937198
The hex bit socket is designed for secure fastening and loosening tasks, featuring a 1/4 in drive for compatibility with various ratchets and drivers. Its 3 mm hex bit ensures precise fit for corresponding fasteners, making it suitable for a range of professional applications.
  • Hex opening securely holds the bit for optimal torque transfer
  • Constructed from heat-treated alloy steel for durability
  • Features a polished chrome finish for corrosion resistance
  • Bit length of 1 in for versatile use
  • Overall length of 1-7/8 in
  • Not insulated
  • Non-sparking