Learn More
MSC Proto Combination Wrench Set, 5/16-2-1/2 Inch, 31 Pc, 1 Set
Catalog No. 501919941
Encompass
This combination wrench set is designed for versatile use across various mechanical applications, offering a comprehensive range of sizes for efficient fastening and loosening tasks.
  • Combination wrench set type
  • Inch system of measurement
  • Size range of 5/16 in–2-1/2 in
  • Tool roll container type
  • Satin finish coating
  • Includes 31 individual wrench sizes
  • Total of 31 pieces
  • Features 12 points for engagement
  • Head offset angle of 15 degrees
  • Lifetime warranty
  • Manufactured in the United States
Catalog No. 50-191-9941 Supplier MSC Supplier No. 38732293
This combination wrench set is designed for versatile use across various mechanical applications, offering a comprehensive range of sizes for efficient fastening and loosening tasks.
  • Combination wrench set type
  • Inch system of measurement
  • Size range of 5/16 in–2-1/2 in
  • Tool roll container type
  • Satin finish coating
  • Includes 31 individual wrench sizes
  • Total of 31 pieces
  • Features 12 points for engagement
  • Head offset angle of 15 degrees
  • Lifetime warranty
  • Manufactured in the United States