Learn More
MSC Proto Carpenter's Tool Box, Steel, 32 in Wide, Brown
Catalog No. 5000534021
Encompass
This carpenter's tool box is constructed from durable steel, designed for organizing and transporting tools. It features a spacious interior with dimensions of 32 in wide, 8.5 in deep, and 9.5 in high, and comes in a classic brown finish. This robust tool storage solution is suitable for various professional and DIY applications.
  • Constructed from steel for durability.
  • Features a spacious interior.
  • Measures 32 in wide, 8.5 in deep, and 9.5 in high.
  • Comes in a brown finish.
  • Does not include drawers.
  • Manufactured in the United States.
Catalog No. 50-005-34021 Supplier MSC Supplier No. 33626151
This carpenter's tool box is constructed from durable steel, designed for organizing and transporting tools. It features a spacious interior with dimensions of 32 in wide, 8.5 in deep, and 9.5 in high, and comes in a classic brown finish. This robust tool storage solution is suitable for various professional and DIY applications.
  • Constructed from steel for durability.
  • Features a spacious interior.
  • Measures 32 in wide, 8.5 in deep, and 9.5 in high.
  • Comes in a brown finish.
  • Does not include drawers.
  • Manufactured in the United States.