Learn More
MSC Proto Combination Wrench, 9/16 in., Satin, 6-Point, 1 ea
Catalog No. 501920042
Encompass
Proto combination wrenches are engineered for professional use, featuring a satin finish for corrosion resistance and a secure grip. The 6-point box end design maximizes contact with fasteners to prevent rounding, making them suitable for demanding industrial and automotive applications.
  • A 6-point box end for increased fastener contact and reduced rounding
  • Satin finish provides enhanced corrosion resistance and a slip-resistant grip
  • Designed with a 15-degree head offset for improved access in tight spaces
  • Tether capable for secure working environments
  • Manufactured from high-quality steel for durability
  • Meets ASME B107.100 standards for performance and safety
Catalog No. 50-192-0042 Supplier MSC Supplier No. 38734737
Proto combination wrenches are engineered for professional use, featuring a satin finish for corrosion resistance and a secure grip. The 6-point box end design maximizes contact with fasteners to prevent rounding, making them suitable for demanding industrial and automotive applications.
  • A 6-point box end for increased fastener contact and reduced rounding
  • Satin finish provides enhanced corrosion resistance and a slip-resistant grip
  • Designed with a 15-degree head offset for improved access in tight spaces
  • Tether capable for secure working environments
  • Manufactured from high-quality steel for durability
  • Meets ASME B107.100 standards for performance and safety