Learn More
MSC Proto Feeler Gage Set, 25 Piece, 0.0015 to 0.04 in Thick, Parallel, Steel
Catalog No. 5000513843
Encompass
This feeler gage set features 25 parallel steel leaves with a thickness range of 0.0015 to 0.04 in. Each leaf measures 12 in long and 1/2 in wide, making it suitable for precise measurements in various applications.
  • Contains 25 leaves
  • Thickness range from 0.0015 to 0.04 in
  • Parallel leaf style for consistent measurements
  • Leaves are 12 in long
  • Leaves are 1/2 in wide
  • Constructed from durable steel
  • Manufactured in the United States
Catalog No. 50-005-13843 Supplier MSC Supplier No. 63940118
This feeler gage set features 25 parallel steel leaves with a thickness range of 0.0015 to 0.04 in. Each leaf measures 12 in long and 1/2 in wide, making it suitable for precise measurements in various applications.
  • Contains 25 leaves
  • Thickness range from 0.0015 to 0.04 in
  • Parallel leaf style for consistent measurements
  • Leaves are 12 in long
  • Leaves are 1/2 in wide
  • Constructed from durable steel
  • Manufactured in the United States