Learn More
MSC Proto Hex Bit Socket, 1/2 Drive, 3/8 Hex, 1 ea
Catalog No. 501920554
Encompass
This Proto hand hex bit socket is engineered for robust performance in various applications. It features a 1/2 inch drive and a 3/8 inch hex bit, making it suitable for driving hex fasteners with precision and durability.
  • Hex bit socket type
  • Drive size: 1/2 in
  • Hex size: 3/8 in
  • Bit length: 1-3/4 in
  • Constructed from chrome alloy steel
  • Stem finish in black oxide
  • Polished chrome finish
  • Overall length of 6-29/32 in
  • Meets GGG-W-641e standards
Catalog No. 50-192-0554 Supplier MSC Supplier No. 60647856
This Proto hand hex bit socket is engineered for robust performance in various applications. It features a 1/2 inch drive and a 3/8 inch hex bit, making it suitable for driving hex fasteners with precision and durability.
  • Hex bit socket type
  • Drive size: 1/2 in
  • Hex size: 3/8 in
  • Bit length: 1-3/4 in
  • Constructed from chrome alloy steel
  • Stem finish in black oxide
  • Polished chrome finish
  • Overall length of 6-29/32 in
  • Meets GGG-W-641e standards