Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(+)-proto-Quercitol 97.0+%, TCI America™
SDP

Catalog No. Q0071100MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
Q0071100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. Q0071100MG Supplier TCI America Supplier No. Q0071100MG

Chemical Identifiers

CAS 488-73-3
Molecular Formula C6H12O5
Molecular Weight (g/mol) 164.157
MDL Number MFCD06797163
InChI Key IMPKVMRTXBRHRB-MBMOQRBOSA-N
Synonym 1L-1,3,4/2,5-Cyclohexanepentol
PubChem CID 441437
ChEBI CHEBI:27371
IUPAC Name (1R,2S,4S,5R)-cyclohexane-1,2,3,4,5-pentol
SMILES C1C(C(C(C(C1O)O)O)O)O

Specifications

Melting Point 237°C
Color White
Quantity 100 mg
Formula Weight 164.16
Percent Purity ≥97.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material (+)-proto-Quercitol
TSCA No
Recommended Storage Freezer
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.