Learn More
Invitrogen™ ProtoArray™ Blocking Buffer Kit
Description
The Blocking Buffer Kit consists of reagents sufficient for preparing Blocking Buffer and Wash Buffer to process 10 ProtoArray™ Human Protein Microarrays for ProtoArray™ protein-protein interaction (PPI), immune response biomarker profiling (IRBP), and antibody specificity profiling (ASP) applications. The kit contains one bottle each of Blocking Buffer and 10X Synthetic Block, and one tube of dithiothreitol (DTT).
Specifications
Specifications
| Type | Blocking Buffer Kit |
| Content And Storage | Store the ProtoArray™ Blocking Buffer Kit at -20°C. |
| For Use With (Equipment) | ProtoArray™ Human Protein Microarrays for immune response biomarker profiling (IRBP), ProtoArray™ Human Protein Microarrays for antibody specificity profiling (ASP), ProtoArray™ Human Protein Microarrays for protein-protein interaction (PPI) |
| Product Line | ProtoArray |
| Quantity | 1 Kit |
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.