Learn More
National Diagnostics PROTOBLUE SAFE 1 LT
SDP
Catalog No. 5089990059
Encompass
ProtoBlue Safe, Eco-friendly, ultra-sensitive colloidal coomassie G-250 stain, 1 LT
Catalog No. 50-899-90059 Supplier National Diagnostics Supplier No. EC7221LTR
ProtoBlue Safe, Eco-friendly, ultra-sensitive colloidal coomassie G-250 stain, 1 LT