Learn More
Sigma Aldrich Fine Chemicals Biosciences Protocatechuc acd phypro100MG
Catalog No. 502902137
Encompass
This substance is a primary reference substance with assigned absolute purity (considering chromatographic purity water residual solvents inorganic impurities). The exact value can be found on the certificate. Produced by PhytoLab GmbH Co. KG
Catalog No. 50-290-2137 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. PHL89766100MG
This substance is a primary reference substance with assigned absolute purity (considering chromatographic purity water residual solvents inorganic impurities). The exact value can be found on the certificate. Produced by PhytoLab GmbH Co. KG