Learn More
Medchemexpress LLC HY-N0294 200mg Medchemexpress, Protocatechuic acid CAS:99-50-3 Purity:>98%
SDP
Catalog No. 501933510
Encompass_Preferred
Medchemexpress, HY-N0294 200mg Protocatechuic acid CAS:99-50-3 Protocatechuic acid is a phenolic compound which exhibits neuroprotective effect. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3510 Supplier Medchemexpress LLC Supplier No. HYN0294200MG
Medchemexpress, HY-N0294 200mg Protocatechuic acid CAS:99-50-3 Protocatechuic acid is a phenolic compound which exhibits neuroprotective effect. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.