Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Synbiosis™ ProtoCOL 3
GREENER_CHOICESDP

Catalog No. 01257310 Shop All Synoptics, Inc. Products
Change view
Click to view available options
Includes:
Integrated 1.4 MP CCD Camera
Integrated 1.4 MP CCD Camera and Mounted PC
Integrated 5 MP CCD Camera
Integrated 5 MP CCD Camera and Mounted PC
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes
01-257-310 Integrated 5 MP CCD Camera
01-257-312 Integrated 1.4 MP CCD Camera
01-257-311 Integrated 1.4 MP CCD Camera and Mounted PC
01-257-309 Integrated 5 MP CCD Camera and Mounted PC
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 01-257-310 Supplier Synbiosis™ Supplier No. PROC3HD
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

ProtoCOL 3 is the next generation instrument for colony counting, zone measurements, and other applications. ProtoCOL 3 has a 3 channel LED lighting system and a 1.4 MP color camera. The HD version has an ultra-HD 5MP camera which captures high definition images.

  • High throughput: ProtoCOL 3 is equipped to be used when large numbers of plates need to be counted or measured rapidly and accurately. All counting and measuring functions are automatic with results instantly sent to the internal reporting system.
  • color imaging: The unique LED lighting in the system gives full color images. The color classification feature of the ProtoCOL 3 software allows for an infinite range of color options on the same plate. A choice of sample holders is included for both light and dark backgrounds, depending on sample type.
  • Wide range of applications: The system can be configured for a wide range of plate types, including pour plates, spiral plates, multi-well, SBA, OPKA, Ames, inhibition zones, AST, and SRD. Plates up to 150 mm can be used on the standard unit. An optional external scanning device can be added should larger plates need to be processed.
  • Extensive report generation: One of the major benefits of an automated system is that reports can be generated within the device. ProtoCOL 3 produces reports that can easily be exported to Excel at the touch of a button.
  • Traceability: All the data generated has a full audit trail. All report generation is traceable and has an audit ‘lock’. Data is CFR compliant and the system can be connected to a LIMS if required.
  • Sensitivity: ProtoCOL 3 uses a highly sensitive CCD camera with a high resolution. Colonies as small as 43 microns (0.043 mm) can be easily counted and zones can be accurately measured to 0.5 mm with a theoretical detection limit of 0.1 mm.

Specifications

Product Type Automated Colony Counter and Zone Measurement System
Material Injection-molded Resin
Model ProtoCOL 3 HD
Outlet Style Standard USA Type B Plug
Product Line Synbiosis
Sample Size 30 to 150 mm
Dimensions (L x W x H) 500 x 400 x 450 mm
Weight (Metric) 20 kg
For Use With (Application) Automatic colony counting and zone measuring
Includes Integrated 5 MP CCD Camera
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.