Learn More
Synbiosis™ ProtoCOL 3 Base Software Package
GREENER_CHOICESDP
Catalog No. 01257320 Shop All Synoptics, Inc. Products
Change view
Click to view available options
For Use With (Application):
Colony counting and zonal analysis
Catalog No. For Use With (Application)
01-257-320 Colony counting and zonal analysis
1 options
Catalog No. 01-257-320 Supplier Synbiosis™ Supplier No. PROC3BASESW
Add to Cart
Add to Cart
Request a Quote
The ProtoCOL 3 Base Software Package includes colony counting and zonal analysis software modules. The colony counting module counts colonies on pour, settle, and spread plates. The inhibition zone measurement software measures the diameter of inhibition zones.
  • Includes colony counting and zonal analysis software modules
  • The colony counting module counts colonies on pour, settle, and spread plates for applications including microbial limit testing, total viable counts, and pour plate dilution series
  • Inhibition zone measurements software module measures the diameter of inhibition zones for applications including Single Radial Immunodiffusion (SRD) plates
    • This module can be combined with UNISTAT™ to determine vaccine potency
Specifications
Product Type Protocol 3 Base Software Package
For Use With (Application) Colony counting and zonal analysis
For Use With (Equipment) ProtoCOL 3 Instruments