Learn More
Synbiosis™ ProtoCOL 3 eAST Software Module for AST (antibiotic susceptibility testing)
GREENER_CHOICESDP
Catalog No. 01257313 Shop All Synoptics, Inc. Products
Change view
Click to view available options
For Use With (Application):
Antibiotic sensitivity plates can be automatically measured and compared to data from all the tested organisms that have breakpoint values in the EUCAST and CLSI databases.
Catalog No. For Use With (Application)
01-257-313 Antibiotic sensitivity plates can be automatically measured and compared to data from all the tested organisms that have breakpoint values in the EUCAST and CLSI databases.
1 options
Catalog No. 01-257-313 Supplier Synbiosis™ Supplier No. PROC3EAST
Add to Cart
Add to Cart
Request a Quote
This ProtoCOL 3 eAST Software Module for AST (antibiotic susceptibility testing) is designed for use with ProtoCOL 3 instruments. The software quickly and simply measures zones around antibiotic discs and calculates antibiotic resistance from results to help guide treatment options.
  • Compare results to data from all the tested organisms that have breakpoint values in the EUCAST and CLSI databases
  • Determine which antibiotics the bacteria are sensitive and resistant to, in less than half the time it takes to perform these tasks manually
  • Generates reports and plate images which can be transferred directly to Excel, eliminating keying and data transfer errors as well as providing a full audit trail
  • Saves hours of time and removes any subjectivity associated with manual zone reading
  • Integrates into GLP and GMP environments with full traceability
Specifications
Product Type ProtoCOL 3 Software Module for AST (antibiotic susceptibility testing)
For Use With (Application) Antibiotic sensitivity plates can be automatically measured and compared to data from all the tested organisms that have breakpoint values in the EUCAST and CLSI databases.
For Use With (Equipment) ProtoCOL 3 Instruments