Learn More
Synbiosis™ ProtoCOL 3 Software Module for Ames
GREENER_CHOICESDP
Catalog No. 01257316 Shop All Synoptics, Inc. Products
Change view
Click to view available options
For Use With (Application):
Automatic measurement of Ames plates
Catalog No. For Use With (Application)
01-257-316 Automatic measurement of Ames plates
1 options
Catalog No. 01-257-316 Supplier Synbiosis™ Supplier No. PROC3AMESMOD
Add to Cart
Add to Cart
The ProtoCOL 3 Software Module for Ames is designed for the automated reading of Ames plates, enabling rapid and precise count, and to determine the mutagenic activity of different chemicals.
  • Designed for the automated reading of Ames plates, enabling rapid and precise count
  • Determine the mutagenic activity of different chemicals,
  • Automatically compare plate results
  • Sold as a single runtime lifetime license
Specifications
Product Type ProtoCOL 3 Software Module for Ames
For Use With (Application) Automatic measurement of Ames plates
For Use With (Equipment) ProtoCOL 3 Instruments