Learn More
Synbiosis™ ProtoCOL 3 Software Module for Chromogenic ID
GREENER_CHOICESDP
Catalog No. 01257322 Shop All Synoptics, Inc. Products
Change view
Click to view available options
For Use With (Application):
Automatic reading of Chromogenic plates
Catalog No. For Use With (Application)
01-257-322 Automatic reading of Chromogenic plates
1 options
Catalog No. 01-257-322 Supplier Synbiosis™ Supplier No. PROC3CHROMEMOD
Add to Cart
Add to Cart
ProtoCOL 3 Software Module for Chromogenic ID is a color recognition module designed to make the analysis of chromogenic agars accurate and simple to interpret.
  • Designed to make the analysis of chromogenic agars accurate and simple to interpret
  • Organisms are identified at the touch of a button with the data and image stored for a permanent electronic record
  • Automatically identify a wide range of microbes cultured on a variety of chromogenic media
  • Automated report generation for traceability
  • Free software upgrades and no annual license fee
Specifications
Product Type ProtoCOL 3 Software Module for Reading Chromogenic Media
For Use With (Application) Automatic reading of Chromogenic plates
For Use With (Equipment) ProtoCOL 3 Instruments