Learn More
Synbiosis™ ProtoCOL 3 Software Module for OPKA (Opsonophagocytic Killing Assay)
GREENER_CHOICESDP
Catalog No. 01257315 Shop All Synoptics, Inc. Products
Change view
Click to view available options
For Use With (Application):
Automatic measurement of OPKA (opsonophagocytic killing assay)
Catalog No. For Use With (Application)
01-257-315 Automatic measurement of OPKA (opsonophagocytic killing assay)
1 options
Catalog No. 01-257-315 Supplier Synbiosis™ Supplier No. PROC3OPKAMOD
Add to Cart
Add to Cart
The ProtoCOL 3 Software Module for OPKA is designed for the automated reading of OPKA plates, enabling rapid and precise count. Count colonies on OPK assay plates to automatically assess the potency of a range of viral vaccines.
  • Designed for the automated reading of OPKA plates, enabling rapid and precise count
  • Automatically assess the potency of a range of viral vaccines
  • Sold as a single runtime lifetime license
Specifications
Product Type ProtoCOL 3 Software Module for OPKA (opsonophagocytic killing assay)
For Use With (Application) Automatic measurement of OPKA (opsonophagocytic killing assay)
For Use With (Equipment) ProtoCOL 3 Instruments