Learn More
Synbiosis™ ProtoCOL 3 Software Module for SBA (serum bactericidal assay)
GREENER_CHOICESDP
Catalog No. 01257318 Shop All Synoptics, Inc. Products
Change view
Click to view available options
For Use With (Application):
Automatic measurement of SBA (serum bactericidal assay)
Catalog No. For Use With (Application)
01-257-318 Automatic measurement of SBA (serum bactericidal assay)
1 options
Catalog No. 01-257-318 Supplier Synbiosis™ Supplier No. PROC3SBAMOD
Add to Cart
Add to Cart
The ProtoCOL 3 Software Module for SBA provides automated reading of serum bacterial assay plates, quickly determining the efficacy of specific bacterial vaccines by counting colonies on SBA plates.
  • Enables quick and easy reading of serum bacterial assay plates, saving analysts vast amounts of time
  • Determines the efficacy of specific bacterial vaccines by counting colonies on SBA plates
  • Sold as a single runtime lifetime license
Specifications
Product Type ProtoCOL 3 Software Module for SBA (serum bactericidal assay)
For Use With (Application) Automatic measurement of SBA (serum bactericidal assay)
For Use With (Equipment) ProtoCOL 3 Instruments