Learn More
Synbiosis™ ProtoCOL 3 Software Module for Sector Plates
GREENER_CHOICESDP
Catalog No. 01257317 Shop All Synoptics, Inc. Products
Change view
Click to view available options
For Use With (Application):
Automatic measurement of multi-sector environmental monitoring plates
Catalog No. For Use With (Application)
01-257-317 Automatic measurement of multi-sector environmental monitoring plates
1 options
Catalog No. 01-257-317 Supplier Synbiosis™ Supplier No. PROC3SECTORMOD
Add to Cart
Add to Cart
The ProtoCOL 3 Software Module for Sector Plates allows the user to count colonies on plates which are divided into different sections, making this module ideal for use with air monitoring plates.
  • Designed for the automated reading of multi-sector plates
  • Ideal for use with air monitoring plates
  • Sold as a single runtime lifetime license
Specifications
Product Type ProtoCOL 3 Software Module for Sector Plates
For Use With (Application) Automatic measurement of multi-sector environmental monitoring plates
For Use With (Equipment) ProtoCOL 3 Instruments