Learn More
Synbiosis™ ProtoCOL 3 Software Module for Spiral Measurement
GREENER_CHOICESDP
Catalog No. 01257314 Shop All Synoptics, Inc. Products
Change view
Click to view available options
For Use With (Application):
Automatic measurement of Spiral plates
Catalog No. For Use With (Application)
01-257-314 Automatic measurement of Spiral plates
1 options
Catalog No. 01-257-314 Supplier Synbiosis™ Supplier No. PROC3SPIRALMOD
Add to Cart
Add to Cart
The ProtoCOL 3 Software Module for Spiral Measurement is a unique application designed to count spiral plates created from major spiral platers including WASP and Eddy Jet spiral platers generating CFU/mL results.
  • Unique application designed to count spiral plates created from major spiral platers, including WASP and Eddy Jet spiral platers generating CFU/mL results
  • Optional software module designed for ProtoCOL 3
  • Spiral plate module is sold as a single runtime lifetime license
Specifications
Product Type ProtoCOL 3 Software Module for Spiral Measurement
For Use With (Application) Automatic measurement of Spiral plates
For Use With (Equipment) ProtoCOL 3 Instruments