Learn More
Protocol™ Decalcifying Solution A, Fisherbrand™
SDP
Catalog No. 11002201 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
32 oz.
1 gal.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-002-201 1 gal.
11-002-202 32 oz.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled
Catalog No. 11-002-201
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled
A gentle decalcifier formulated from semiconductor grade hydrochloric acid, EDTA, and highly purified reagent grade water.
  • Can be used to safely decalcify all types of tissue samples from needle biopsies to cortical bone
  • Mild enough that most samples can be left in overnight
Specifications
Product Type Decalcifying Solution
Quantity 1 gal.
Solution Type Decalcifier
Form Solution
Made in the USA