Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Protocol™ Formalin, Fisherbrand™
SDP

Catalog No. 11002205 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
32 oz.
1 gal.
2.5 gal.
5 gal.
55 gal.
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-002-205 1 gal.
11-002-206 2.5 gal.
11-002-207 5 gal.
11-002-208 55 gal.
11-002-229 32 oz.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled
Catalog No. 11-002-205
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled

Ready-to-use buffered formalin in bulk packaging.

  • 10% neutral buffered formalin fixative
  • Used to preserve the cellular structure and morphology of tissue for examination under a microscope by preventing decay and maintaining its molecular integrity during the tissue processing steps for staining and analysis
  • Featuring a pH level of 7.0

Specifications

For Use With (Equipment) All Tissue Processors
Grade Histological
Product Type Formalin
Quantity 1 gal.
Solution Type Formalin
Form Solution

Made in the USA

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.