Learn More
Leica Biosystems PROTOCOL HRS HEMATOXYLN 1GAL
Catalog No. NC0096440
Encompass_Preferred
Harris Hematoxylin produces a deep, purple-blue stain. It is generally used in regressive methods, but may also be used progressively both in cytology and histology preparations. Harris Hematoxylin does not contain mercury.
Catalog No. NC0096440 Supplier Leica Biosystems Supplier No. 3801560
Harris Hematoxylin produces a deep, purple-blue stain. It is generally used in regressive methods, but may also be used progressively both in cytology and histology preparations. Harris Hematoxylin does not contain mercury.