Learn More
Description
- Formaldehyde-free solution provides the morphological detail of a standard aldehyde fixative while preserving high levels of immunoreactivity.
- A glyoxal based formulation
Specifications
Specifications
| For Use With (Equipment) | All Tissue Processors |
| Grade | Histological |
| Product Type | Fixative |
| Quantity | 1 gal. |
| Solution Type | Fixative |
| Form | Solution |
Made in the USA
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
