Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Protocol™ SecureMount, Fisherbrand™
SDP

Catalog No. 11002235 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
16 oz.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-002-235 16 oz.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled
Catalog No. 11-002-235
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled

A rapid-drying, toluene-based mounting medium/liquid coverglass.

  • Compatible with xylene, and xylene-substitute clearing agents including d-Limonene
  • Slides mounted with Acrymount Plus may be screened in as little as 10 minutes
  • Formulated to reduce oxidation, preventing yellowing and fading in H and E, IHC and special stains after coverslipping
  • Maintains an optimal refractive index for viewing and photo microscopy
  • For labs seeking to eliminate final clearing with xylene, tissue may be coverslipped directly from absolute Isopropyl Alcohol
  • Compatible with both manual and most automated coverslipping processes.

Specifications

Quantity 16 oz.
Product Type Mounting Medium
Reagent Type Mounting Solution
Volume (Metric) 473 mL

Made in the USA

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.