Learn More
Description
- Enhances the quality of immunochemical staining
- Minimizes antigen crosslinking
Specifications
Specifications
| Chemical Name or Material | PROTOCOL™ Zinc Formalin |
| Quantity | 1 gal. |
| Synonym | dihydrogen oxide, dihydrogen monoxide |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.