Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PROTOCOL™ Zinc Formalin, Fisher Healthcare™

Catalog No. 23313096 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
1 gal.
5 gal.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-313096 5 gal.
23-313095 1 gal.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 23-313096

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

  • Enhances the quality of immunochemical staining
  • Minimizes antigen crosslinking

Specifications

Chemical Name or Material PROTOCOL™ Zinc Formalin
Quantity 5 gal.
Synonym dihydrogen oxide, dihydrogen monoxide
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.