Learn More
Cayman Chemical ProtodIoscIn 10mg
SDP
Catalog No. 502952772
Encompass_Preferred
A natural steroidal saponin; cytotoxic to a range of cancer cells in vitro, including leukemia, colon cancer, and prostate cancer cells (GI50 ≤ 2 µM)
Catalog No. 50-295-2772 Supplier Cayman Chemical Supplier No. 1188710 MG
A natural steroidal saponin; cytotoxic to a range of cancer cells in vitro, including leukemia, colon cancer, and prostate cancer cells (GI50 ≤ 2 µM)