Learn More
Cayman Chemical Protodioscin, Purity: ≥95%, CAS: 55056-80-9, 50 mg
SDP
Catalog No. 502952271
Encompass_Preferred
A natural steroidal saponin; cytotoxic to a range of cancer cells in vitro, including leukemia, colon cancer, and prostate cancer cells (GI50 ≤ 2 µM)
Catalog No. 50-295-2271 Supplier Cayman Chemical Supplier No. 1188750 MG
A natural steroidal saponin; cytotoxic to a range of cancer cells in vitro, including leukemia, colon cancer, and prostate cancer cells (GI50 ≤ 2 µM)