Learn More
AdipoGen Protodioscin
SDP
Catalog No. 502560194
Encompass_Preferred
Chemical. CAS 55056-80-9. Formula C51H84O22. MW 1049.2. Protodioscin is a steroidal saponin first isolated from plants, with anticancer, antihyperlipidemic, antioxidant, anti-inflammatory and neuroprotective properties. It displays cytotoxic, apoptosis incuding and antiproliferative activity against a range of cancer cells. Protodioscin has also been shown to have aphrodisiac properties.
Catalog No. 50-256-0194 Supplier AdipoGen Supplier No. CDXP0451M010
Chemical. CAS 55056-80-9. Formula C51H84O22. MW 1049.2. Protodioscin is a steroidal saponin first isolated from plants, with anticancer, antihyperlipidemic, antioxidant, anti-inflammatory and neuroprotective properties. It displays cytotoxic, apoptosis incuding and antiproliferative activity against a range of cancer cells. Protodioscin has also been shown to have aphrodisiac properties.