Learn More
National Diagnostics ProtoGel (30%), 37.5:1 Acrylamide to Bisacrylamide Stabilized Solution, 1 liter
SDP
Catalog No. 5089990118
Encompass
ProtoGel (30%); National Diagnostics; 37.5:1 acrylamide to bisacrylamide stabilized soln.; Optimized for SDS-PAGE of proteins (Laemmli gels); Consistently crystal clear gels, zero fluorescence; Stabilized for long shelf life; Size: 1L
Catalog No. 50-899-90118 Supplier National Diagnostics Supplier No. EC8901LTR
ProtoGel (30%); National Diagnostics; 37.5:1 acrylamide to bisacrylamide stabilized soln.; Optimized for SDS-PAGE of proteins (Laemmli gels); Consistently crystal clear gels, zero fluorescence; Stabilized for long shelf life; Size: 1L