Learn More
National Diagnostics PROTOGEL (30%) 450 ML
SDP
Catalog No. 5089990119
Encompass
ProtoGel (30%), 30% concentrated solution of acrylamide and bis-acrylamide, 37.5 : 1 ratio. Filtered, deionized, and stabilized, 450 ML
Catalog No. 50-899-90119 Supplier National Diagnostics Supplier No. EC890450ML
ProtoGel (30%), 30% concentrated solution of acrylamide and bis-acrylamide, 37.5 : 1 ratio. Filtered, deionized, and stabilized, 450 ML