Learn More
National Diagnostics PROTOGEL (40%) 1 LT
SDP
Catalog No. 5089990120
Encompass
ProtoGel (40%), Concentrated solution of acrylamide and bis-acrylamide, 37.5 : 1 ratio. Filtered, deionized, and stabilized, 1 LT
Catalog No. 50-899-90120 Supplier National Diagnostics Supplier No. EC8911LTR
ProtoGel (40%), Concentrated solution of acrylamide and bis-acrylamide, 37.5 : 1 ratio. Filtered, deionized, and stabilized, 1 LT