Learn More
National Diagnostics PROTOGEL (40%) 450 ML
SDP
Catalog No. 5089990121
Encompass
ProtoGel (40%), Concentrated solution of acrylamide and bis-acrylamide, 37.5 : 1 ratio. Filtered, deionized, and stabilized, 450 ML
Catalog No. 50-899-90121 Supplier National Diagnostics Supplier No. EC891450ML
ProtoGel (40%), Concentrated solution of acrylamide and bis-acrylamide, 37.5 : 1 ratio. Filtered, deionized, and stabilized, 450 ML