Learn More
National Diagnostics PROTOGEL QUICK-CAST 12% 450 ML
SDP
Catalog No. 5089990126
Encompass
ProtoGel Quick-Cast 12%, Ready-to-use gel solution for SDS-PAGE that combines the convenience of pre-cast gels with the economy of casting your own, 450 ML
Catalog No. 50-899-90126 Supplier National Diagnostics Supplier No. EC895450ML
ProtoGel Quick-Cast 12%, Ready-to-use gel solution for SDS-PAGE that combines the convenience of pre-cast gels with the economy of casting your own, 450 ML