Learn More
National Diagnostics ProtoGel Resolving Buffer (4X) 450ML
SDP
Catalog No. 5089990123
Encompass
Traditional Laemmli Resolving Gel Buffer (4X). Formulated with 0.2 Micron Filtration.Ultra-Pure Reagents in 18 MegOhm Water.Crystal Clear, Reproducible Gels. Catalog number: EC-892. The use of National Diagnostics' ProtoGel Resolving Buffer will ensure the purity and performance of your Laemmli gels. When diluted as directed ProtoGel Resolving Buffer forms a gel of 0.375 M Tris-HCl and 0.1% SDS, pH 8.8. Storage: ProtoGel Resolving Buffer is stable for 24 months when stored tightly capped in a dark area at room temperature.
Catalog No. 50-899-90123 Supplier National Diagnostics Supplier No. EC892450ML
Add to Cart
Add to Cart
Traditional Laemmli Resolving Gel Buffer (4X). Formulated with 0.2 Micron Filtration.Ultra-Pure Reagents in 18 MegOhm Water.Crystal Clear, Reproducible Gels. Catalog number: EC-892. The use of National Diagnostics' ProtoGel Resolving Buffer will ensure the purity and performance of your Laemmli gels. When diluted as directed ProtoGel Resolving Buffer forms a gel of 0.375 M Tris-HCl and 0.1% SDS, pH 8.8. Storage: ProtoGel Resolving Buffer is stable for 24 months when stored tightly capped in a dark area at room temperature.