Learn More
National Diagnostics PROTOGEL STACKING BUFFER 200 M
SDP
Catalog No. 5089990124
Encompass
ProtoGel Stacking Buffer, 4X Laemmli Tris-HCl stacking gel buffer for clear, reproducible SDS-PAGE, 200 ML
Catalog No. 50-899-90124 Supplier National Diagnostics Supplier No. EC893200ML
Add to Cart
Add to Cart
ProtoGel Stacking Buffer, 4X Laemmli Tris-HCl stacking gel buffer for clear, reproducible SDS-PAGE, 200 ML